Learn More
Medchemexpress LLC N-hexanoyl-DL-homoserine lactone | 106983-28-2 | 98.0% | 199.25 | C10H17NO3 | 25 MG
SDP
Catalog No. 5000209592
Encompass_Preferred
N-hexanoyl-DL-homoserine lactone is an N-acyl homoserine lactone (AHL) quorum sensing signaling molecule used in research to study bacterial communication and related biological effects.
  • Used to study quorum sensing and bacterial signaling.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Reported purity 98.0%.
  • Molecular formula C10H17NO3 and molecular weight 199.25.
  • Provided in multiple packaging sizes including 25 mg.
Catalog No. 50-002-09592 Supplier Medchemexpress LLC Supplier No. 25MGHYW045071
N-hexanoyl-DL-homoserine lactone is an N-acyl homoserine lactone (AHL) quorum sensing signaling molecule used in research to study bacterial communication and related biological effects.
  • Used to study quorum sensing and bacterial signaling.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Reported purity 98.0%.
  • Molecular formula C10H17NO3 and molecular weight 199.25.
  • Provided in multiple packaging sizes including 25 mg.