Learn More
Medchemexpress LLC N-hexanoyl-L-homoserine lactone | 147852-83-3 | MFCD12912260 | 98.8% | 199.25 g/mol | C10H17NO3 | 5 MG

Supplier: Medchemexpress LLC HY133685R5MG
N-hexanoyl-L-homoserine lactone is a short-chain acyl-homoserine lactone used as a bacterial quorum-sensing signaling molecule and as an analytical standard for microbiology and biochemical research. Provided as a purified standard, it is suitable for assay development, calibration, and mechanistic studies.
- Used as a quorum-sensing standard in bacterial signaling and biofilm research.
- High purity (98.8%) for analytical and research applications.
- Molecular formula C10H17NO3; molecular weight 199.25 g/mol.
- Available in small solid quantities and preprepared DMSO solutions for convenience.
- Certificate of analysis provided for lot traceability.
- Soluble in common organic solvents such as DMSO.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.