Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-hexanoyl-L-homoserine lactone | 147852-83-3 | MFCD12912260 | 98.8% | C10H17NO3 | 10MG
SDP

Catalog No. 5000260423
Encompass_Preferred

N-hexanoyl-L-homoserine lactone is a short-chain N-acyl homoserine lactone commonly used as a bacterial quorum-sensing signaling molecule in research. It serves as a high-purity chemical standard for studies of intercellular communication and related biochemical assays.

  • Purity 98.8% suitable for analytical and biological assays.
  • Molecular weight 199.25 g/mol supports stoichiometric calculations.
  • Used to study quorum sensing and signaling in gram-negative bacteria.
  • Supplied in multiple small sizes for flexibility in experimental design.

Catalog No. 50-002-60423 Supplier Medchemexpress LLC Supplier No. HY133685R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-hexanoyl-L-homoserine lactone is a short-chain N-acyl homoserine lactone commonly used as a bacterial quorum-sensing signaling molecule in research. It serves as a high-purity chemical standard for studies of intercellular communication and related biochemical assays.

  • Purity 98.8% suitable for analytical and biological assays.
  • Molecular weight 199.25 g/mol supports stoichiometric calculations.
  • Used to study quorum sensing and signaling in gram-negative bacteria.
  • Supplied in multiple small sizes for flexibility in experimental design.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.