Learn More
Medchemexpress LLC N-hexanoylsphingosine | 124753-97-5 | 99.8% | 397.63 | C24H47NO3 | 10 MG
SDP
Catalog No. 5000294852
Encompass_Preferred
C6 ceramide is a short-chain, cell-permeable ceramide pathway activator used in cell-based research to induce apoptosis and modulate signaling pathways relevant to cancer progression and chemosensitivity.
  • Induces apoptosis and cell cycle arrest in cancer cells.
  • Enhances chemosensitivity in drug-resistant models.
  • Soluble in DMSO at high concentrations for in vitro use.
  • High purity for reproducible experimental results.
  • Stable when stored as a powder at recommended temperatures.
  • Available in multiple pack sizes for experimental flexibility.
Catalog No. 50-002-94852 Supplier Medchemexpress LLC Supplier No. HY1954210MG
C6 ceramide is a short-chain, cell-permeable ceramide pathway activator used in cell-based research to induce apoptosis and modulate signaling pathways relevant to cancer progression and chemosensitivity.
  • Induces apoptosis and cell cycle arrest in cancer cells.
  • Enhances chemosensitivity in drug-resistant models.
  • Soluble in DMSO at high concentrations for in vitro use.
  • High purity for reproducible experimental results.
  • Stable when stored as a powder at recommended temperatures.
  • Available in multiple pack sizes for experimental flexibility.