Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexyl formate, 97%, Thermo Scientific™

Catalog No. AAL0622630
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0622630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0622630 Supplier Thermo Scientific Chemicals Supplier No. L0622630

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-33-4
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00046146
InChI Key OUGPMNMLWKSBRI-UHFFFAOYSA-N
Synonym n-hexyl formate, formic acid, hexyl ester, n-hexyl methanoate, hexyl methanoate, formic acid hexyl ester, hexyl formiate, unii-9gpu7pgv1g, fema no. 2570, 9gpu7pgv1g, hexylformate
PubChem CID 61177
IUPAC Name hexyl formate
SMILES CCCCCCOC=O

Specifications

Melting Point -63°C
Density 0.875
Boiling Point 155°C
Flash Point 37°C (98°F)
Refractive Index 1.408
Quantity 250 g
UN Number UN3272
Beilstein 1701629
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material n-Hexyl formate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 211-087-1
RTECSNumber LQ8595000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.