Learn More
Strem, An Ascensus Company CAS# 4721-24-8. 1g. n-Hexylphosphonic acid, min. 97% HPA. MFCD00013961
SDP
Catalog No. 5090110240
Encompass
CAS# 4721-24-8. 1g. n-Hexylphosphonic acid, min. 97% HPA. MFCD00013961. Molecular Weight: 166.16. Molecular Formula: CH3(CH2)5P(O)(OH)2. Color/form: white to off-white pwdr. Strem# 15-2410. http://www.strem.com/catalog/v/15-2410/
Catalog No. 50-901-10240 Supplier Strem, An Ascensus Company Supplier No. 1524101G
CAS# 4721-24-8. 1g. n-Hexylphosphonic acid, min. 97% HPA. MFCD00013961. Molecular Weight: 166.16. Molecular Formula: CH3(CH2)5P(O)(OH)2. Color/form: white to off-white pwdr. Strem# 15-2410. http://www.strem.com/catalog/v/15-2410/