Learn More
Cayman Chemical N-HIppuryl-HIs-Leuhydrate 50mg
SDP
Catalog No. 502964309
Encompass_Preferred
A synthetic ACE substrate; has been used in the in vitro identification of ACE inhibitors
Catalog No. 50-296-4309 Supplier Cayman Chemical Supplier No. 2332950 MG
A synthetic ACE substrate; has been used in the in vitro identification of ACE inhibitors