Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxy-1,8-naphthalimide, 97%, Thermo Scientific™

Catalog No. AAB2248522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2248522 100 g
AAB2248514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2248522 Supplier Thermo Scientific Chemicals Supplier No. B2248522

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in chemical research & chemical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7797-81-1
Molecular Formula C12H7NO3
Molecular Weight (g/mol) 213.192
MDL Number MFCD00065062
InChI Key KTWCUGUUDHJVIH-UHFFFAOYSA-N
Synonym n-hydroxynaphthalimide, n-hydroxy-1,8-naphthalimide, naphthalimide, n-hydroxy, naphthalhydroxamic acid, 2-hydroxy-1h-benzo de isoquinoline-1,3 2h-dione, n-hydroxy-1,8-naphthalenedicarboximide, 2-hydroxy-1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxybenzo de isoquinoline-1,3-dione, 1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxy, unii-dzg9avw5v2
PubChem CID 82263
IUPAC Name 2-hydroxybenzo[de]isoquinoline-1,3-dione
SMILES C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)O

Specifications

Melting Point 283°C to 286°C
Quantity 100 g
Beilstein 178595
Solubility Information Insoluble in water.
Formula Weight 213.19
Percent Purity 97%
Chemical Name or Material N-Hydroxy-1,8-naphthalimide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 232-251-9
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.