Learn More
Medchemexpress LLC N-hydroxy-2,2-dimethyl-N-(phenylmethyl)-butanamide | 1956370-21-0 | MFCD30738006 | 99.9% | 221.29 g/mol | C13H19NO2 | 200 MG
SDP
Catalog No. 5000213081
Encompass_Preferred
RIPA-56 is a potent, selective, and metabolically stable inhibitor of receptor-interacting protein 1 (RIP1) kinase intended for research use. Supplied as a solid with high reported purity and characterized enzymatic and cellular potency, it is used in preclinical cellular assays and animal studies to probe RIP1-dependent signaling and inflammatory pathways.
  • Potent, selective RIP1 kinase inhibition with low-nanomolar activity.
  • High purity suitable for biochemical and cell-based assays.
  • Available in multiple sizes and solution formats for flexible dosing.
  • Solid, white to off-white appearance for easy handling.
  • Suitable for preclinical cellular and in vivo model studies.
Catalog No. 50-002-13081 Supplier Medchemexpress LLC Supplier No. HY101032200MG
RIPA-56 is a potent, selective, and metabolically stable inhibitor of receptor-interacting protein 1 (RIP1) kinase intended for research use. Supplied as a solid with high reported purity and characterized enzymatic and cellular potency, it is used in preclinical cellular assays and animal studies to probe RIP1-dependent signaling and inflammatory pathways.
  • Potent, selective RIP1 kinase inhibition with low-nanomolar activity.
  • High purity suitable for biochemical and cell-based assays.
  • Available in multiple sizes and solution formats for flexible dosing.
  • Solid, white to off-white appearance for easy handling.
  • Suitable for preclinical cellular and in vivo model studies.