Learn More
Medchemexpress LLC N-hydroxy-3-[1-(phenylsulfanylmethyl)triazol-4-yl]benzamide | 1316652-41-1 | MFCD21363002 | 99.3% | 326.37 g/mol | C16H14N4O2S | 100 MG
SDP
Catalog No. 5000227588
Encompass_Preferred
NCC-149 (CAS 1316652-41-1) is a selective histone deacetylase 8 (HDAC8) inhibitor used in neural differentiation and other HDAC8-targeted research. The compound has molecular formula C16H14N4O2S and a molecular weight of 326.37 g/mol. Certificate of analysis lists purity ~99.3%. Store powder at -20°C for long-term stability. For research use only.
  • Selective HDAC8 inhibitor suitable for target validation and neural differentiation assays.
  • Validated molecular identity: C16H14N4O2S; MW 326.37 g/mol.
  • High purity reported by COA (≈99.3%).
  • Available in multiple pack sizes, including 100 MG.
  • Powder storage at -20°C extends shelf life; follow SDS for solvent storage recommendations.
  • Supporting documentation available: SDS and COA for handling and quality verification.
Catalog No. 50-002-27588 Supplier Medchemexpress LLC Supplier No. HY117348100MG
NCC-149 (CAS 1316652-41-1) is a selective histone deacetylase 8 (HDAC8) inhibitor used in neural differentiation and other HDAC8-targeted research. The compound has molecular formula C16H14N4O2S and a molecular weight of 326.37 g/mol. Certificate of analysis lists purity ~99.3%. Store powder at -20°C for long-term stability. For research use only.
  • Selective HDAC8 inhibitor suitable for target validation and neural differentiation assays.
  • Validated molecular identity: C16H14N4O2S; MW 326.37 g/mol.
  • High purity reported by COA (≈99.3%).
  • Available in multiple pack sizes, including 100 MG.
  • Powder storage at -20°C extends shelf life; follow SDS for solvent storage recommendations.
  • Supporting documentation available: SDS and COA for handling and quality verification.