Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxy-3-[1-(phenylthio)methyl-1H-1,2,3-triazol-4-yl]benzamide 96.0+%, TCI America™
SDP

Catalog No. H13405MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13405MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H13405MG Supplier TCI America Supplier No. H13405MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1316652-41-1
Molecular Formula C16H14N4O2S
Molecular Weight (g/mol) 326.37
MDL Number MFCD21363002
InChI Key YMCCQRYCIRCIGS-UHFFFAOYSA-N
Synonym NCC-149
PubChem CID 53327227
IUPAC Name N-oxo-3-{1-[(phenylsulfanyl)methyl]-2,3-dihydro-1H-1,2,3-triazol-4-yl}benzamide
SMILES O=NC(=O)C1=CC=CC(=C1)C1=CN(CSC2=CC=CC=C2)NN1

Specifications

Melting Point 153°C
Quantity 5 mg
Formula Weight 326.37
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Hydroxy-3-[1-(phenylthio)methyl-1H-1,2,3-triazol-4-yl]benzamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.