Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N'-Hydroxy-3-methyl-2-pyridinecarboximidamide, 97%, Thermo Scientific™

Catalog No. MO01289DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO01289DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01289DA Supplier Thermo Scientific Chemicals Supplier No. MO01289DA
Add to Cart

Chemical Identifiers

CAS 690632-33-8
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.169
MDL Number MFCD06200877
InChI Key YIBWTWSINGSDFJ-UHFFFAOYSA-N
Synonym 3-methylpyridine-2-amidoxime, n'-hydroxy-3-methyl-2-pyridinecarboximidamide, n'-hydroxy-3-methylpyridine-2-carboximidamide, z-n'-hydroxy-3-methylpyridine-2-carboximidamide, uqdfgzccscdnso-uhfffaoysa-n, n-hydroxy-3-methyl-pyridine-2-carboxamidine, 3-methyl-1h-pyridin-2-ylidene-nitrosomethanamine, 3-methyl-1h-pyridin-2-ylidene nitroso methanamine
PubChem CID 6816486
IUPAC Name (3-methyl-1H-pyridin-2-ylidene)-nitrosomethanamine
SMILES CC1=CC=CNC1=C(N)N=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material N'-Hydroxy-3-methyl-2-pyridinecarboximidamide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.