Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-hydroxy-3-pyridinecarboximidamide | 1594-58-7 | MFCD00125875; MFCD00265955 | 97.0% | 137.14 | C6H7N3O | 10 G
SDP

Catalog No. 5000300300
Encompass_Preferred

3-Pyridylamide oxime (N-hydroxy-3-pyridinecarboximidamide) is a small-molecule synthetic intermediate commonly used in pharmaceutical and medicinal chemistry for preparation of target analogues and downstream intermediates. It is supplied as a stable solid intended for research and organic synthesis applications.

  • High purity: 97.0% (NMR).
  • Molecular formula C6H7N3O; molecular weight 137.14.
  • Appearance: white to light yellow solid.
  • Storage: powder stable at -20°C (3 years) or 4°C (2 years).
  • Available in small pack sizes suitable for laboratory synthesis (10 g).

Catalog No. 50-003-00300 Supplier Medchemexpress LLC Supplier No. HYW01580510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Pyridylamide oxime (N-hydroxy-3-pyridinecarboximidamide) is a small-molecule synthetic intermediate commonly used in pharmaceutical and medicinal chemistry for preparation of target analogues and downstream intermediates. It is supplied as a stable solid intended for research and organic synthesis applications.

  • High purity: 97.0% (NMR).
  • Molecular formula C6H7N3O; molecular weight 137.14.
  • Appearance: white to light yellow solid.
  • Storage: powder stable at -20°C (3 years) or 4°C (2 years).
  • Available in small pack sizes suitable for laboratory synthesis (10 g).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.