Learn More
Medchemexpress LLC N-hydroxy-4-[[(2-methoxyphenyl)carbamoylamino]methyl]benzamide | 1454902-97-6 | 95.1% | 315.32 g·mol⁻¹ | C16H17N3O4 | 5 MG
SDP
Catalog No. 5000221448
Encompass_Preferred
Elevenostat (JB3-22) is a small-molecule selective histone deacetylase 11 (HDAC11) inhibitor for research use. Reported HDAC11 potency is IC50 ≈ 0.235 μM. It has been shown to induce apoptosis in multiple myeloma cell lines and to affect mouse oocyte maturation. Provided as a purified solid for biochemical and cellular studies; molecular formula C16H17N3O4, molecular weight 315.32 g·mol⁻¹.
  • Selective HDAC11 inhibition (IC50 ≈ 0.235 μM).
  • Induces apoptosis in multiple myeloma cell lines.
  • Affects mouse oocyte maturation in preclinical studies.
  • Supplied as a purified research-grade solid.
  • Suitable for biochemical and cellular assays.
  • Soluble in DMSO for typical assay preparations.
Catalog No. 50-002-21448 Supplier Medchemexpress LLC Supplier No. HY1457575MG
Elevenostat (JB3-22) is a small-molecule selective histone deacetylase 11 (HDAC11) inhibitor for research use. Reported HDAC11 potency is IC50 ≈ 0.235 μM. It has been shown to induce apoptosis in multiple myeloma cell lines and to affect mouse oocyte maturation. Provided as a purified solid for biochemical and cellular studies; molecular formula C16H17N3O4, molecular weight 315.32 g·mol⁻¹.
  • Selective HDAC11 inhibition (IC50 ≈ 0.235 μM).
  • Induces apoptosis in multiple myeloma cell lines.
  • Affects mouse oocyte maturation in preclinical studies.
  • Supplied as a purified research-grade solid.
  • Suitable for biochemical and cellular assays.
  • Soluble in DMSO for typical assay preparations.