Learn More
Medchemexpress LLC N-hydroxy-4-[[(2S)-3-methyl-2-phenylbutanoyl]amino]benzamide | 935881-37-1 | MFCD17676151 | 98.5% | 312.36 g/mol | C18H20N2O3 | 5 MG
SDP
Catalog No. 5000262968
Encompass_Preferred
AR-42 is a potent pan-histone deacetylase (HDAC) inhibitor used in preclinical research to study epigenetic regulation, acetylation status, and related signaling pathways. It induces growth inhibition, cell-cycle arrest, and apoptosis in multiple cancer cell lines and is supplied as a purified research chemical for in vitro and in vivo studies.
  • Potent pan-HDAC inhibitor with reported IC50 approximately 16 nM.
  • Orally bioavailable and active in preclinical models.
  • Induces growth inhibition, cell-cycle arrest, and apoptosis.
  • Promotes hyperacetylation of histones H3 and H4, and α-tubulin.
  • Supplied as a high-purity research chemical (~98.5%).
  • Soluble in common organic solvents such as DMSO and ethanol for assay preparation.
Catalog No. 50-002-62968 Supplier Medchemexpress LLC Supplier No. HY132655MG
AR-42 is a potent pan-histone deacetylase (HDAC) inhibitor used in preclinical research to study epigenetic regulation, acetylation status, and related signaling pathways. It induces growth inhibition, cell-cycle arrest, and apoptosis in multiple cancer cell lines and is supplied as a purified research chemical for in vitro and in vivo studies.
  • Potent pan-HDAC inhibitor with reported IC50 approximately 16 nM.
  • Orally bioavailable and active in preclinical models.
  • Induces growth inhibition, cell-cycle arrest, and apoptosis.
  • Promotes hyperacetylation of histones H3 and H4, and α-tubulin.
  • Supplied as a high-purity research chemical (~98.5%).
  • Soluble in common organic solvents such as DMSO and ethanol for assay preparation.