Learn More
Medchemexpress LLC N-hydroxy-4-((3-(2-methoxyphenyl)ureido)methyl)benzamide | 1454902-97-6 | 95.1% | C16H17N3O4 | 10MG
SDP
Catalog No. 5000258682
Encompass_Preferred
Elevenostat (JB3-22) is a small-molecule selective inhibitor of histone deacetylase 11 (HDAC11) used in biochemical, cellular, and preclinical in vivo research. It inhibits HDAC11 activity (reported IC50 = 0.235 μM), induces apoptosis in multiple myeloma cells, and has demonstrated antitumor effects in mouse models. The compound is supplied as a powder or as a 10 mM solution in DMSO at research-grade purity and includes recommended storage conditions for stability.
  • Selective HDAC11 inhibition with reported IC50 of 0.235 μM.
  • Induces apoptosis in multiple myeloma cells and shows in vivo antitumor activity.
  • Affects oocyte maturation in cellular assays, enabling reproductive biology studies.
  • Available as solid and 10 mM DMSO solution for assay flexibility.
  • Stable under recommended storage conditions for long-term use.
  • Provided at research-grade purity suitable for biochemical and cellular studies.
Catalog No. 50-002-58682 Supplier Medchemexpress LLC Supplier No. HY14575710MG
Elevenostat (JB3-22) is a small-molecule selective inhibitor of histone deacetylase 11 (HDAC11) used in biochemical, cellular, and preclinical in vivo research. It inhibits HDAC11 activity (reported IC50 = 0.235 μM), induces apoptosis in multiple myeloma cells, and has demonstrated antitumor effects in mouse models. The compound is supplied as a powder or as a 10 mM solution in DMSO at research-grade purity and includes recommended storage conditions for stability.
  • Selective HDAC11 inhibition with reported IC50 of 0.235 μM.
  • Induces apoptosis in multiple myeloma cells and shows in vivo antitumor activity.
  • Affects oocyte maturation in cellular assays, enabling reproductive biology studies.
  • Available as solid and 10 mM DMSO solution for assay flexibility.
  • Stable under recommended storage conditions for long-term use.
  • Provided at research-grade purity suitable for biochemical and cellular studies.