Learn More
Medchemexpress LLC N-hydroxy-4-(3-(pyridin-2-yl)phenyl)benzamide | 2151853-97-1 | MFCD32063565 | 99.8% | 293.32 g/mol | C17H15N3O2 | 100MG
SDP
Catalog No. 5000235407
Encompass_Preferred
MPT0G211 is a potent, orally active, selective histone deacetylase 6 (HDAC6) inhibitor provided for research use. It exhibits >1,000-fold selectivity for HDAC6, a reported IC50 of 0.291 nM, and is reported to penetrate the blood-brain barrier. Preclinical studies report modulation of tau phosphorylation, improvement of cognitive deficits in Alzheimer disease models, and anti-metastatic, neuroprotective, and anticancer activities.
  • Potent HDAC6 inhibition (IC50 = 0.291 nM).
  • Greater than 1,000-fold selectivity for HDAC6 over other isoforms.
  • Reported blood-brain barrier penetration for central nervous system studies.
  • Demonstrated activity in Alzheimer disease and anticancer models.
  • High chemical purity (reported 99.8%).
  • Available in multiple solid and solution pack sizes for flexible use.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-35407 Supplier Medchemexpress LLC Supplier No. HY123976100MG
MPT0G211 is a potent, orally active, selective histone deacetylase 6 (HDAC6) inhibitor provided for research use. It exhibits >1,000-fold selectivity for HDAC6, a reported IC50 of 0.291 nM, and is reported to penetrate the blood-brain barrier. Preclinical studies report modulation of tau phosphorylation, improvement of cognitive deficits in Alzheimer disease models, and anti-metastatic, neuroprotective, and anticancer activities.
  • Potent HDAC6 inhibition (IC50 = 0.291 nM).
  • Greater than 1,000-fold selectivity for HDAC6 over other isoforms.
  • Reported blood-brain barrier penetration for central nervous system studies.
  • Demonstrated activity in Alzheimer disease and anticancer models.
  • High chemical purity (reported 99.8%).
  • Available in multiple solid and solution pack sizes for flexible use.
  • For research use only; not for human or veterinary use.