Learn More
Medchemexpress LLC N-hydroxy-4-((quinolin-8-ylamino)methyl)benzamide | 2151853-97-1 | 99.8% | 293.32 g·mol-1 | C17H15N3O2 | 10MG
SDP
Catalog No. 5000235390
Encompass_Preferred
MPT0G211 is a potent, orally active, and selective HDAC6 inhibitor intended for preclinical research applications. It is used in biochemical and cellular studies to probe HDAC6-related pathways relevant to neurodegeneration and oncology, and is characterized by sub-nanomolar potency and high chemical purity.
  • Selective HDAC6 inhibition (IC50 0.291 nM).
  • Sub-nanomolar potency suitable for biochemical assays.
  • High purity, 99.8%.
  • Suitable for in vitro and in vivo preclinical studies.
  • Soluble in DMSO for stock solution preparation.
  • Available in small research pack sizes for screening and confirmation studies.
Catalog No. 50-002-35390 Supplier Medchemexpress LLC Supplier No. HY12397610MG
MPT0G211 is a potent, orally active, and selective HDAC6 inhibitor intended for preclinical research applications. It is used in biochemical and cellular studies to probe HDAC6-related pathways relevant to neurodegeneration and oncology, and is characterized by sub-nanomolar potency and high chemical purity.
  • Selective HDAC6 inhibition (IC50 0.291 nM).
  • Sub-nanomolar potency suitable for biochemical assays.
  • High purity, 99.8%.
  • Suitable for in vitro and in vivo preclinical studies.
  • Soluble in DMSO for stock solution preparation.
  • Available in small research pack sizes for screening and confirmation studies.