Learn More
Medchemexpress LLC N-hydroxy-4-((quinolin-8-ylamino)methyl)benzamide | 2151853-97-1 | 99.8% | 293.32 g/mol | C17H15N3O2 | 25MG
SDP
Catalog No. 5000236755
Encompass_Preferred
MPT0G211 is a potent, orally active, and selective histone deacetylase 6 (HDAC6) inhibitor used for preclinical research. It exhibits sub-nanomolar potency against HDAC6 (IC50 = 0.291 nM), strong selectivity over other HDAC isoforms, and is reported to penetrate the blood-brain barrier. Reported activities include reduction of tau phosphorylation and improvement of cognitive deficits in Alzheimer's disease models, along with anti-metastatic, neuroprotective, and anticancer effects.
  • Potent HDAC6 inhibition (IC50 = 0.291 nM)
  • >1,000-fold selectivity for HDAC6 versus other HDAC isoforms
  • Orally active with reported blood-brain barrier penetration
  • Demonstrated reduction of tau phosphorylation in disease models
  • Reported cognitive benefit in Alzheimer's disease model studies
  • High reported purity suitable for research use (≈99.8%)
  • Available in small research pack sizes for in vitro and in vivo studies
Catalog No. 50-002-36755 Supplier Medchemexpress LLC Supplier No. HY12397625MG
MPT0G211 is a potent, orally active, and selective histone deacetylase 6 (HDAC6) inhibitor used for preclinical research. It exhibits sub-nanomolar potency against HDAC6 (IC50 = 0.291 nM), strong selectivity over other HDAC isoforms, and is reported to penetrate the blood-brain barrier. Reported activities include reduction of tau phosphorylation and improvement of cognitive deficits in Alzheimer's disease models, along with anti-metastatic, neuroprotective, and anticancer effects.
  • Potent HDAC6 inhibition (IC50 = 0.291 nM)
  • >1,000-fold selectivity for HDAC6 versus other HDAC isoforms
  • Orally active with reported blood-brain barrier penetration
  • Demonstrated reduction of tau phosphorylation in disease models
  • Reported cognitive benefit in Alzheimer's disease model studies
  • High reported purity suitable for research use (≈99.8%)
  • Available in small research pack sizes for in vitro and in vivo studies