Learn More
Medchemexpress LLC N-hydroxy-7-(naphthalen-2-ylsulfanyl)heptanamide | 926908-04-5 | MFCD10568146 | 99.8% | 303.42 g/mol | C17H21NO2S | 25 MG
SDP
Catalog No. 5000228060
Encompass_Preferred
HNHA is a small-molecule histone deacetylase (HDAC) inhibitor used for biomedical research. It inhibits HDACs with reported IC50 around 100 nM, arrests the cell cycle at G1/S via p21 induction, and has demonstrated antitumor and anti-angiogenic activity in preclinical studies.
  • Potent HDAC inhibitor with reported IC50 of ~100 nM.
  • High purity, listed as 99.76% on the product page.
  • Molecular formula C17H21NO2S and molecular weight 303.42 g/mol.
  • Provided in small research quantities, including 25 mg packages.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-28060 Supplier Medchemexpress LLC Supplier No. HY11867225MG
HNHA is a small-molecule histone deacetylase (HDAC) inhibitor used for biomedical research. It inhibits HDACs with reported IC50 around 100 nM, arrests the cell cycle at G1/S via p21 induction, and has demonstrated antitumor and anti-angiogenic activity in preclinical studies.
  • Potent HDAC inhibitor with reported IC50 of ~100 nM.
  • High purity, listed as 99.76% on the product page.
  • Molecular formula C17H21NO2S and molecular weight 303.42 g/mol.
  • Provided in small research quantities, including 25 mg packages.
  • For research use only; not for human or diagnostic use.