Learn More
Medchemexpress LLC Nω-Hydroxy-nor-l-arginine acetate | 1140844-63-8 | 99.5% | 296.28 | 10 MM * 1 ML
SDP
Catalog No. 5000439594
Encompass_Preferred
nor-NOHA acetate is a selective and reversible Arginase inhibitor. It induces Apoptosis in ARG2-expressing cells under hypoxia and exhibits anti-leukemic activity. Additionally, nor-NOHA acetate improves liver ischemia-reperfusion injury and can be utilized in studies related to tuberculosis, endothelial dysfunction, immunosuppression, and metabolism.
  • Selective and reversible arginase inhibitor.
  • Induces apoptosis in ARG2-expressing cells under hypoxia.
  • Exhibits anti-leukemic activity.
  • Improves liver ischemia-reperfusion injury.
  • Can be used in studies of tuberculosis, endothelial dysfunction, immunosuppression, and metabolism.
Catalog No. 50-004-39594 Supplier Medchemexpress LLC Supplier No. HY112885A10MM/1ML
nor-NOHA acetate is a selective and reversible Arginase inhibitor. It induces Apoptosis in ARG2-expressing cells under hypoxia and exhibits anti-leukemic activity. Additionally, nor-NOHA acetate improves liver ischemia-reperfusion injury and can be utilized in studies related to tuberculosis, endothelial dysfunction, immunosuppression, and metabolism.
  • Selective and reversible arginase inhibitor.
  • Induces apoptosis in ARG2-expressing cells under hypoxia.
  • Exhibits anti-leukemic activity.
  • Improves liver ischemia-reperfusion injury.
  • Can be used in studies of tuberculosis, endothelial dysfunction, immunosuppression, and metabolism.