Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxybenzenecarboximidamide, 97%, Thermo Scientific™

Catalog No. RF01827DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RF01827DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RF01827DA Supplier Thermo Scientific Chemicals Supplier No. RF01827DA
Add to Cart

Chemical Identifiers

CAS 613-92-3
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00031485 MFCD00474011
InChI Key MXOQNVMDKHLYCZ-UHFFFAOYSA-N
Synonym benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim
PubChem CID 7259353
ChEBI CHEBI:83354
SMILES N\C(=N/O)C1=CC=CC=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula C6H5C(NH2)=NOH
Quantity 1 g
IUPAC Name N'-hydroxybenzenecarboximidamide
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material N'-Hydroxybenzenecarboximidamide
Hazard Category Warning
Hazard Statement GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.