Learn More
Medchemexpress LLC N-hydroxybenzenesulfonamide | 599-71-3 | ≥98.0% | 1 G
SDP
Catalog No. 5000232800
Encompass_Preferred
Piloty's acid (N-hydroxybenzenesulfonamide) is a nitroxyl (HNO) donor used in chemical and biological research. It decomposes under basic conditions to release HNO and the benzenesulfinate anion, and is supplied in small laboratory pack sizes for synthesis and assay work.
  • Nitroxyl (HNO) donor used in chemical and biological studies.
  • White crystalline solid with a melting point around 186 °C.
  • Molecular formula C6H7NO3S, molecular weight 173.2 g·mol⁻¹.
  • High purity (≥98%) suitable for research applications.
  • Available in small laboratory pack sizes for routine synthesis and assays.
Catalog No. 50-002-32800 Supplier Medchemexpress LLC Supplier No. HY1209901G
Add to Cart
Add to Cart
Piloty's acid (N-hydroxybenzenesulfonamide) is a nitroxyl (HNO) donor used in chemical and biological research. It decomposes under basic conditions to release HNO and the benzenesulfinate anion, and is supplied in small laboratory pack sizes for synthesis and assay work.
  • Nitroxyl (HNO) donor used in chemical and biological studies.
  • White crystalline solid with a melting point around 186 °C.
  • Molecular formula C6H7NO3S, molecular weight 173.2 g·mol⁻¹.
  • High purity (≥98%) suitable for research applications.
  • Available in small laboratory pack sizes for routine synthesis and assays.