Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-hydroxyhydantoin carbamate | 2331255-53-7 | 98.5% | 100 MG
SDP

Catalog No. 5000230428
Encompass_Preferred

ABC99 is an N-hydroxyhydantoin carbamate small-molecule inhibitor of the Wnt-deacylating enzyme NOTUM (reported IC50 = 13 nM). It preserves Wnt3A signaling in cell-based assays and is supplied as a powder with recommended cold storage.

  • Selective NOTUM inhibitor (IC50 = 13 nM).
  • N-hydroxyhydantoin carbamate scaffold.
  • Molecular formula C22H21ClN4O5.
  • Molecular weight 456.88.
  • Powder form; store powder at -20°C (3 years) or 4°C (2 years).
  • Soluble in DMSO: 25 mg/mL (≈54.7 mM) with heating and sonication.
  • Preserves Wnt3A signaling in cell-based assays.
  • Available as a 100 mg research quantity.

Catalog No. 50-002-30428 Supplier Medchemexpress LLC Supplier No. HY122832100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ABC99 is an N-hydroxyhydantoin carbamate small-molecule inhibitor of the Wnt-deacylating enzyme NOTUM (reported IC50 = 13 nM). It preserves Wnt3A signaling in cell-based assays and is supplied as a powder with recommended cold storage.

  • Selective NOTUM inhibitor (IC50 = 13 nM).
  • N-hydroxyhydantoin carbamate scaffold.
  • Molecular formula C22H21ClN4O5.
  • Molecular weight 456.88.
  • Powder form; store powder at -20°C (3 years) or 4°C (2 years).
  • Soluble in DMSO: 25 mg/mL (≈54.7 mM) with heating and sonication.
  • Preserves Wnt3A signaling in cell-based assays.
  • Available as a 100 mg research quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.