Learn More
Medchemexpress LLC N-hydroxyisonicotinamidine | 1594-57-6 | MFCD00125873 | ≥95.0% | 137.14 g/mol | C6H7N3O | 25 G
SDP
Catalog No. 5000303491
Encompass_Preferred
N-Hydroxyisonicotinamidine is an amidoxime-based chemical intermediate supplied as a powder for research use. It functions as a collector and a building block in organic synthesis, notably in the preparation of substituted α-1,2,4-oxadiazole esters. The compound has formula C6H7N3O and a molar mass of 137.14 g/mol, and is soluble in DMSO at high concentrations.
Catalog No. 50-003-03491 Supplier Medchemexpress LLC Supplier No. 25GHYW023757
N-Hydroxyisonicotinamidine is an amidoxime-based chemical intermediate supplied as a powder for research use. It functions as a collector and a building block in organic synthesis, notably in the preparation of substituted α-1,2,4-oxadiazole esters. The compound has formula C6H7N3O and a molar mass of 137.14 g/mol, and is soluble in DMSO at high concentrations.