Learn More
Medchemexpress LLC N-hydroxyisonicotinamidine | 1594-57-6 | MFCD00125873 | 99.9% | 137.14 g·mol^-1 | C6H7N3O | 10 G
SDP
Catalog No. 5000300122
Encompass_Preferred
N-hydroxyisonicotinamidine is an amidoxime-based chemical intermediate and collector used in research-scale organic synthesis. It is employed in the preparation of substituted α-1,2,4-oxadiazole esters and related heterocycles. The compound has formula C6H7N3O and a molecular weight of 137.14, and is supplied as a high-purity reagent for laboratory use.
  • Amidoxime functional group suitable for heterocycle construction.
  • Documented application in synthesis of substituted α-1,2,4-oxadiazole esters.
  • Provided as a high-purity reagent for research applications.
  • Stable as a powder when stored under recommended conditions.
  • Available in small pack sizes tailored for laboratory use.
Catalog No. 50-003-00122 Supplier Medchemexpress LLC Supplier No. HYW02375710G
N-hydroxyisonicotinamidine is an amidoxime-based chemical intermediate and collector used in research-scale organic synthesis. It is employed in the preparation of substituted α-1,2,4-oxadiazole esters and related heterocycles. The compound has formula C6H7N3O and a molecular weight of 137.14, and is supplied as a high-purity reagent for laboratory use.
  • Amidoxime functional group suitable for heterocycle construction.
  • Documented application in synthesis of substituted α-1,2,4-oxadiazole esters.
  • Provided as a high-purity reagent for research applications.
  • Stable as a powder when stored under recommended conditions.
  • Available in small pack sizes tailored for laboratory use.