Learn More
Medchemexpress LLC N-hydroxyisonicotinamidine | 1594-57-6 | MFCD00125873 | 99.9% | 137.14 g/mol | C6H7N3O | 5 G
SDP
Catalog No. 5000301453
Encompass_Preferred
N-hydroxyisonicotinamidine is an amidoxime-based collector and chemical intermediate used in organic synthesis. It is commonly employed as a precursor in the preparation of substituted α-1,2,4-oxadiazole esters and related heterocycles. Supplied for research use only, consult the product datasheet for handling and storage details.
  • Amidoxime-based collector for organic synthesis.
  • Useful precursor for substituted α-1,2,4-oxadiazole esters.
  • Molecular formula C6H7N3O; molecular weight 137.14 g/mol.
  • Stable as a powder under low-temperature storage; follow datasheet storage conditions.
  • For research use only; not validated for clinical or therapeutic use.
Catalog No. 50-003-01453 Supplier Medchemexpress LLC Supplier No. HYW0237575G
N-hydroxyisonicotinamidine is an amidoxime-based collector and chemical intermediate used in organic synthesis. It is commonly employed as a precursor in the preparation of substituted α-1,2,4-oxadiazole esters and related heterocycles. Supplied for research use only, consult the product datasheet for handling and storage details.
  • Amidoxime-based collector for organic synthesis.
  • Useful precursor for substituted α-1,2,4-oxadiazole esters.
  • Molecular formula C6H7N3O; molecular weight 137.14 g/mol.
  • Stable as a powder under low-temperature storage; follow datasheet storage conditions.
  • For research use only; not validated for clinical or therapeutic use.