Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-hydroxymethyl succinimide | 5146-68-9 | MFCD01328751 | 95.0% | 129.11 g/mol | C5H7NO3 | 5 G
SDP

Catalog No. 5000281887
Encompass_Preferred

N-hydroxymethyl succinimide is a promoiety used as a prodrug carrier for non-steroidal anti-inflammatory drugs (NSAIDs). It enhances the lipophilicity of parent compounds and can reduce gastrointestinal toxicity, making it suitable for research into anti-inflammatory drug delivery systems.

  • Used as a prodrug promoiety for NSAIDs
  • Enhances lipophilicity of parent drugs
  • May reduce gastrointestinal toxicity in formulations
  • Chemical formula C5H7NO3; molecular weight 129.11 g/mol
  • CAS number 5146-68-9
  • Powder storage: -20°C (3 years), 4°C (2 years)

Catalog No. 50-002-81887 Supplier Medchemexpress LLC Supplier No. HYW1601275G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-hydroxymethyl succinimide is a promoiety used as a prodrug carrier for non-steroidal anti-inflammatory drugs (NSAIDs). It enhances the lipophilicity of parent compounds and can reduce gastrointestinal toxicity, making it suitable for research into anti-inflammatory drug delivery systems.

  • Used as a prodrug promoiety for NSAIDs
  • Enhances lipophilicity of parent drugs
  • May reduce gastrointestinal toxicity in formulations
  • Chemical formula C5H7NO3; molecular weight 129.11 g/mol
  • CAS number 5146-68-9
  • Powder storage: -20°C (3 years), 4°C (2 years)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.