Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Hydroxymethylnicot 100g | 3569-99-1 | 152.15 g/mol | C7H8N2O2 | 100 G
SDP

Catalog No. 5000227321
Encompass_Preferred

N-(Hydroxymethyl)nicotinamide (CAS 3569-99-1) is a nicotinamide derivative described as an antimicrobial agent. Supplied as a solid for research use, it has a defined molecular formula and high reported purity suitable for laboratory studies.

  • Antimicrobial agent.
  • Molecular weight 152.15 g/mol.
  • Chemical formula C7H8N2O2.
  • Purity 99.82%.
  • Soluble in DMSO at 100 mg/mL (ultrasonic recommended).
  • Available as a solid and as a DMSO solution for in vitro use.
  • For research use only.

Catalog No. 50-002-27321 Supplier Medchemexpress LLC Supplier No. HY116993100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-(Hydroxymethyl)nicotinamide (CAS 3569-99-1) is a nicotinamide derivative described as an antimicrobial agent. Supplied as a solid for research use, it has a defined molecular formula and high reported purity suitable for laboratory studies.

  • Antimicrobial agent.
  • Molecular weight 152.15 g/mol.
  • Chemical formula C7H8N2O2.
  • Purity 99.82%.
  • Soluble in DMSO at 100 mg/mL (ultrasonic recommended).
  • Available as a solid and as a DMSO solution for in vitro use.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.