Learn More
Medchemexpress LLC N-Hydroxymethylnicot 10mM 1mL | 3569-99-1 | 1 ML
SDP
Catalog No. 5000229216
Encompass_Preferred
N-(Hydroxymethyl)nicotinamide is an antimicrobic small-molecule reagent provided as a ready-to-use 10 mM solution (1 mL in DMSO) and as solid material across multiple pack sizes. The compound has molecular formula C7H8N2O2, a molecular weight of 152.15 g/mol, and a reported purity of 99.82%. Recommended storage conditions differ for powder and solvent preparations.
Catalog No. 50-002-29216 Supplier Medchemexpress LLC Supplier No. HY11699310MM 1ML
N-(Hydroxymethyl)nicotinamide is an antimicrobic small-molecule reagent provided as a ready-to-use 10 mM solution (1 mL in DMSO) and as solid material across multiple pack sizes. The compound has molecular formula C7H8N2O2, a molecular weight of 152.15 g/mol, and a reported purity of 99.82%. Recommended storage conditions differ for powder and solvent preparations.