Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Hydroxymethylnicot 25g | 3569-99-1 | 152.15 g/mol | C7H8N2O2 | 25 G
SDP

Catalog No. 5000307607
Encompass_Preferred

N-(Hydroxymethyl)nicotinamide (CAS 3569-99-1) is a nicotinamide derivative used as an antimicrobial research chemical. It is supplied as a white to off-white solid powder with high purity and defined storage conditions to maintain stability. Offered in multiple pack sizes and solution formats for laboratory use.

  • High purity: 99.82%.
  • Molecular formula C7H8N2O2; molecular weight 152.15 g/mol.
  • Physical state: white to off-white solid powder.
  • Available pack sizes include 25 g and solution (10 mM in DMSO).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).

Catalog No. 50-003-07607 Supplier Medchemexpress LLC Supplier No. HY11699325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-(Hydroxymethyl)nicotinamide (CAS 3569-99-1) is a nicotinamide derivative used as an antimicrobial research chemical. It is supplied as a white to off-white solid powder with high purity and defined storage conditions to maintain stability. Offered in multiple pack sizes and solution formats for laboratory use.

  • High purity: 99.82%.
  • Molecular formula C7H8N2O2; molecular weight 152.15 g/mol.
  • Physical state: white to off-white solid powder.
  • Available pack sizes include 25 g and solution (10 mM in DMSO).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.