Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxynaphthalimide 97%, Thermo Scientific™

Catalog No. AC412080250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC412080250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412080250 Supplier Thermo Scientific Chemicals Supplier No. 412080250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7797-81-1
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C12H7NO3
Quantity 25g
Beilstein 21, V, 361
Synonym n-hydroxynaphthalimide, n-hydroxy-1,8-naphthalimide, naphthalimide, n-hydroxy, naphthalhydroxamic acid, 2-hydroxy-1h-benzo de isoquinoline-1,3 2h-dione, n-hydroxy-1,8-naphthalenedicarboximide, 2-hydroxy-1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxybenzo de isoquinoline-1,3-dione, 1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxy, unii-dzg9avw5v2
InChI Key KTWCUGUUDHJVIH-UHFFFAOYSA-N
SMILES C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)O
IUPAC Name 2-hydroxybenzo[de]isoquinoline-1,3-dione
Molecular Weight (g/mol) 213.192
PubChem CID 82263
Formula Weight 213.19
Chemical Name or Material N-Hydroxynaphthalimide, 97%
Show More Show Less
EINECSNumber 232-251-9
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.