Learn More
Medchemexpress LLC N-hydroxyphthalimide | 524-38-9 | MFCD00005891 | 99.6% | 163.13 | C8H5NO3 | 500g
SDP
Catalog No. 5000269263
Encompass_Preferred
N-Hydroxyphthalimide is a blocking agent and catalyst N-Hydroxyphthalimide promotes oxidation reactions by generating PINO free radicals and activating hydrogen atom transfer processes N-Hydroxyphthalimide reduces the expression of anti-apoptotic proteins Survivin and Bcl-xL and activates caspase 9 and caspase 3 N-Hydroxyphthalimide induces Apoptosis N-Hydroxyphthalimide inhibits the phosphorylation of mTOR (Ser2448 Ser2481) and Akt (Ser473) N-Hydroxyphthalimide has anticancer effects against breast and colon cancer[1][2][3]
Catalog No. 50-002-69263 Supplier Medchemexpress LLC Supplier No. HYY0396500G
N-Hydroxyphthalimide is a blocking agent and catalyst N-Hydroxyphthalimide promotes oxidation reactions by generating PINO free radicals and activating hydrogen atom transfer processes N-Hydroxyphthalimide reduces the expression of anti-apoptotic proteins Survivin and Bcl-xL and activates caspase 9 and caspase 3 N-Hydroxyphthalimide induces Apoptosis N-Hydroxyphthalimide inhibits the phosphorylation of mTOR (Ser2448 Ser2481) and Akt (Ser473) N-Hydroxyphthalimide has anticancer effects against breast and colon cancer[1][2][3]