Learn More
Medchemexpress LLC N-Hydroxypipecolic acid | 115819-92-6 | 250 MG
SDP
Catalog No. 5000421987
Encompass_Preferred
N-Hydroxypipecolic acid is a plant metabolite and a systemic acquired resistance (SAR) regulator. It orchestrates SAR establishment in concert with the immune signal salicylic acid. This compound accumulates systemically in plant foliage in response to pathogen attack and induces SAR to bacterial and oomycete infection.
  • Direct induction of SAR gene expression
  • Signal amplification
  • Priming for enhanced defense activation
  • Positive interplay with salicylic acid signaling to ensure elevated plant immunity
  • Flavin-dependent-monooxygenase1 (FMO1) functions downstream of Pip by hydroxylating Pip to generate the product.
Catalog No. 50-004-21987 Supplier Medchemexpress LLC Supplier No. 250MGHYN7378
N-Hydroxypipecolic acid is a plant metabolite and a systemic acquired resistance (SAR) regulator. It orchestrates SAR establishment in concert with the immune signal salicylic acid. This compound accumulates systemically in plant foliage in response to pathogen attack and induces SAR to bacterial and oomycete infection.
  • Direct induction of SAR gene expression
  • Signal amplification
  • Priming for enhanced defense activation
  • Positive interplay with salicylic acid signaling to ensure elevated plant immunity
  • Flavin-dependent-monooxygenase1 (FMO1) functions downstream of Pip by hydroxylating Pip to generate the product.