Learn More
Medchemexpress LLC HY-N7378 5mg Medchemexpress, N-Hydroxypipecolic acid CAS:115819-92-6 Purity:>98%
SDP
Catalog No. 502031627
Encompass_Preferred
Medchemexpress, HY-N7378 5mg N-Hydroxypipecolic acid CAS:115819-92-6 N-Hydroxypipecolic acid (1-Hydroxy-2-piperidinecarboxylic acid), a plant metabolite and a systemic acquired resistance (SAR) regulator, orchestrates SAR establishment in concert with the immune signal salicylic acid. N-Hydroxypipecolic acid accumulates systemically in the plant foliage in response to pathogen attack. N-Hydroxypipecolic acid induces SAR to bacterial and oomycete infection[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1627 Supplier Medchemexpress LLC Supplier No. HYN73785MG
Medchemexpress, HY-N7378 5mg N-Hydroxypipecolic acid CAS:115819-92-6 N-Hydroxypipecolic acid (1-Hydroxy-2-piperidinecarboxylic acid), a plant metabolite and a systemic acquired resistance (SAR) regulator, orchestrates SAR establishment in concert with the immune signal salicylic acid. N-Hydroxypipecolic acid accumulates systemically in the plant foliage in response to pathogen attack. N-Hydroxypipecolic acid induces SAR to bacterial and oomycete infection[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.