Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-hydroxysuccinimide-activated PEG4-Val-Cit-PAB-MMAE | 2762518-86-3 | MFCD09835251 | 98.9% | 1496.78 | C74H117N11O21 | 5 MG
SDP

Catalog No. 5000203398
Encompass_Preferred

OSu-PEG4-VC-PAB-MMAE is an N-hydroxysuccinimide (OSu) activated PEG4 val-cit-para-aminobenzyl (PAB) linker conjugated to monomethylauristatin E (MMAE). It is supplied as a white to off-white solid for research use in antibody-drug conjugate synthesis and site-specific conjugation.

  • Suitable for site-specific conjugation in ADC construction.
  • Contains a cleavable val-cit linker and self-immolative PAB spacer.
  • Provided as a high-purity solid, facilitating storage and handling.
  • Stable when stored at -80°C and protected from light under inert atmosphere.
  • Soluble in common organic solvents and DMSO for reaction preparation.

Catalog No. 50-002-03398 Supplier Medchemexpress LLC Supplier No. HY1484265MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OSu-PEG4-VC-PAB-MMAE is an N-hydroxysuccinimide (OSu) activated PEG4 val-cit-para-aminobenzyl (PAB) linker conjugated to monomethylauristatin E (MMAE). It is supplied as a white to off-white solid for research use in antibody-drug conjugate synthesis and site-specific conjugation.

  • Suitable for site-specific conjugation in ADC construction.
  • Contains a cleavable val-cit linker and self-immolative PAB spacer.
  • Provided as a high-purity solid, facilitating storage and handling.
  • Stable when stored at -80°C and protected from light under inert atmosphere.
  • Soluble in common organic solvents and DMSO for reaction preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.