Learn More
Abcam N-Hydro x ysuccinimide, Amine-reactive compound, 5G
Catalog No. 502706724
Encompass_Preferred
MW 115.09 Da, Purity >99%. Amine-reactive compound. Used to modify, label and crosslink amine groups. Tool used for protein-protein interactions, N-terminal residue identification and the location of phosphorylated or O-glycosylated residues.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6724 Supplier Abcam Supplier No. AB1412755G
MW 115.09 Da, Purity >99%. Amine-reactive compound. Used to modify, label and crosslink amine groups. Tool used for protein-protein interactions, N-terminal residue identification and the location of phosphorylated or O-glycosylated residues.
The product is subject to the following: Abcam Restricted Use Statement