Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxysuccinimide, Spectrum™ Chemical
SDP

Catalog No. 18620869 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-869 25 g
18-620-870 100 g
18-620-871 250 g
18-620-872 1 kg
18-620-873 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-869 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H249525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Synonym: 1-Hydroxypyrrolidine-2,5-Dione, HOSu, 1-Hydroxysuccinimide

N-Hydroxysuccinimide is an organic compound often found in biochemistry and used as an activating reagent for carboxylic acids. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 6066-82-6

Specifications

Quantity 25 g
Molecular Weight (g/mol) 115.09
Percent Purity 98%
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material N-Hydroxysuccinimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.