Learn More
Medchemexpress LLC HY-W002213 500mg Medchemexpress, N-Hydroxysulfosuccinimide (sodium) CAS:106627-54-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW002213500MG
Encompass_Preferred
Medchemexpress, HY-W002213 500mg N-Hydroxysulfosuccinimide (sodium) CAS:106627-54-7 N-Hydroxysulfosuccinimide (sodium) is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3210