Learn More
Cayman Chemical N-Hydroxysulfosuccinimide so
SDP
Supplier:  Cayman Chemical 20680500 MG
Encompass_Preferred
A modifying reagent that, in the presence of a cross-linking reagent, converts carboxyl groups to amine-reactive sulfo-NHS esters
Catalog No. 50-298-4966