Learn More
Cayman Chemical N-Hydroxysulfosuccinimide so
SDP
Catalog No. 502981753
Encompass_Preferred
A modifying reagent that, in the presence of a cross-linking reagent, converts carboxyl groups to amine-reactive sulfo-NHS esters
Catalog No. 50-298-1753 Supplier Cayman Chemical Supplier No. 20680250 MG
A modifying reagent that, in the presence of a cross-linking reagent, converts carboxyl groups to amine-reactive sulfo-NHS esters