Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-hydroxysulfosuccinimide (sodium) | 106627-54-7 | 100.0% | 217.13 | C4H4NNaO6S | 1 ML
SDP

Catalog No. 5000202363
Encompass_Preferred

N-Hydroxysulfosuccinimide (sodium) is a sulfonated N-hydroxysuccinimide reagent used to activate carboxyl groups for coupling to primary amines in bioconjugation and antibody-drug conjugate synthesis. It increases aqueous solubility compared with NHS esters and is provided as a ready-to-use DMSO solution for research applications.

  • cas 106627-54-7
  • molecular formula C4H4NNaO6S
  • molecular weight 217.13
  • purity 99.99% (manufacturer reported)
  • form solid (white to off-white); supplied as 10 mM solution in DMSO, 1 mL
  • soluble in DMSO; various in vivo vehicle solubilities reported
  • storage sealed, away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month)
  • intended for research use only (bioconjugation/adc linker)

Catalog No. 50-002-02363 Supplier Medchemexpress LLC Supplier No. HYW00221310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Hydroxysulfosuccinimide (sodium) is a sulfonated N-hydroxysuccinimide reagent used to activate carboxyl groups for coupling to primary amines in bioconjugation and antibody-drug conjugate synthesis. It increases aqueous solubility compared with NHS esters and is provided as a ready-to-use DMSO solution for research applications.

  • cas 106627-54-7
  • molecular formula C4H4NNaO6S
  • molecular weight 217.13
  • purity 99.99% (manufacturer reported)
  • form solid (white to off-white); supplied as 10 mM solution in DMSO, 1 mL
  • soluble in DMSO; various in vivo vehicle solubilities reported
  • storage sealed, away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month)
  • intended for research use only (bioconjugation/adc linker)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.