Learn More
Medchemexpress LLC N-hydroxysulfosuccinimide (sodium) | 106627-54-7 | 100.0% | 217.13 | C4H4NNaO6S | 25 G
SDP
Supplier:  Medchemexpress LLC HYW00221325G
Encompass_Preferred
N-hydroxysulfosuccinimide (sodium) is the water-soluble sulfonated analogue of N-hydroxysuccinimide used to activate carboxyl groups to form amine-reactive esters for bioconjugation and linker chemistry. It is commonly used in protein modification and antibody-drug conjugate synthesis workflows, and is supplied as a solid or as a DMSO solution.
  • Water-soluble sulfonated NHS reagent for bioconjugation.
  • Activates carboxyl groups to form amine-reactive esters.
  • Available as solid and as 10 mM solution in DMSO.
  • High reported purity suitable for research applications.
Catalog No. 50-002-04223