Learn More
Medchemexpress LLC N-hydroxysulfosuccinimide sodium | 106627-54-7 | MFCD00043100 | 100.0% | 217.13 g/mol | C4H4NNaO6S | 10 G
SDP
Catalog No. 5000203533
Encompass_Preferred
N-hydroxysulfosuccinimide sodium is a water-soluble N-hydroxysuccinimide derivative used to activate carboxyl groups for conjugation to primary amines, enabling formation of stable amide bonds for bioconjugation and linker synthesis. It is supplied as a solid and in solution forms for research applications.
  • Water-soluble activated ester for amine coupling.
  • Reacts with carboxyl groups to form stable amide bonds.
  • Commonly used in protein and peptide bioconjugation.
  • Available in multiple pack sizes, including 10 g.
  • High purity suitable for research applications.
Catalog No. 50-002-03533 Supplier Medchemexpress LLC Supplier No. HYW00221310G
N-hydroxysulfosuccinimide sodium is a water-soluble N-hydroxysuccinimide derivative used to activate carboxyl groups for conjugation to primary amines, enabling formation of stable amide bonds for bioconjugation and linker synthesis. It is supplied as a solid and in solution forms for research applications.
  • Water-soluble activated ester for amine coupling.
  • Reacts with carboxyl groups to form stable amide bonds.
  • Commonly used in protein and peptide bioconjugation.
  • Available in multiple pack sizes, including 10 g.
  • High purity suitable for research applications.