Learn More
Medchemexpress LLC N-hydroxysulfosuccinimide (sodium) | 106627-54-7 | MFCD00043100 | 100.0% | 217.13 g/mol | C4H4NNaO6S | 100 G
SDP
Catalog No. 5000202165
Encompass_Preferred
N-hydroxysulfosuccinimide (sodium) is the sodium salt of sulfo-N-hydroxysuccinimide, supplied as a white to off-white solid. It serves as a water-soluble activated ester reagent for preparing amine-reactive sulfo-NHS esters for protein modification and bioconjugation, and is used as a non-cleavable linker in antibody-drug conjugate synthesis. Store sealed and protected from moisture.
  • Water-soluble activated ester for efficient amine coupling.
  • Suitable for protein modification and bioconjugation.
  • Used as a non-cleavable linker in antibody-drug conjugate synthesis.
  • High purity (99.99%).
  • White to off-white solid, stable when stored sealed and dry at 4°C.
  • Molecular weight 217.13 g/mol; formula C4H4NNaO6S.
  • Available in solid pack sizes up to 100 g.
Catalog No. 50-002-02165 Supplier Medchemexpress LLC Supplier No. HYW002213100G
N-hydroxysulfosuccinimide (sodium) is the sodium salt of sulfo-N-hydroxysuccinimide, supplied as a white to off-white solid. It serves as a water-soluble activated ester reagent for preparing amine-reactive sulfo-NHS esters for protein modification and bioconjugation, and is used as a non-cleavable linker in antibody-drug conjugate synthesis. Store sealed and protected from moisture.
  • Water-soluble activated ester for efficient amine coupling.
  • Suitable for protein modification and bioconjugation.
  • Used as a non-cleavable linker in antibody-drug conjugate synthesis.
  • High purity (99.99%).
  • White to off-white solid, stable when stored sealed and dry at 4°C.
  • Molecular weight 217.13 g/mol; formula C4H4NNaO6S.
  • Available in solid pack sizes up to 100 g.