Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Hydroxysulfosuccinimide Sodium Salt

Catalog No. 564855G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-485-5G 5 g
56-485-1G 1 g
56-485-250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 56-485-5G Supplier Sigma Aldrich Supplier No. 564855G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Hydroxysulfosuccinimide sodium salt is a water-soluble sulfonated analog of N-hydroxysuccinimide. It can undergo a coupling reaction with carboxylic acids in the presence of carbodiimides such as 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide(EDC) to form hydrophilic N-hydroxysulfosuccinimide active esters. These esters are useful in protein cross-linking experiments.Sulfo-NHS is used for preparing hydrophilic active esters, e.g. for crosslinking agents

Specifications

CAS 106627-54-7
Molecular Formula C4H4NNaO6S
Linear Formula C4H4NNaO6S
MDL Number MFCD00043100
Quantity 5 g
Synonym Hydroxy-2,5-dioxopyrrolidine-3-sulfonicacid sodium salt; Sulfo-NHS
Molecular Weight (g/mol) 217.13 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.