Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxysulfosuccinimide sodium salt, 97%

Catalog No. p-7035089
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52795MD 250 mg
AAH5279503 1 g
AAH5279506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52795MD Supplier Thermo Scientific Chemicals Supplier No. H52795MD
Only null left
Add to Cart

CAS: 106627-54-7 | C4H4NNaO6S | 217.127 g/mol

N-Hydroxysulfosuccinimide sodium salt is used as crosslinking agents for preparing hydrophilic active esters. It is also used to activate carboxylic acids to the more reactive succinimidyl esters for coupling, biotinylation and conjugation with fluorophores. Further, it acts as a hydrophilic ligand and also used in covalently binding proteins to carboxyl containing beads, resins, other nanoparticles. It is involved in the conversion of carboxyl groups to primary amine-reactive sulfo-NHS esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Hydroxysulfosuccinimide sodium salt is used as crosslinking agents for preparing hydrophilic active esters. It is also used to activate carboxylic acids to the more reactive succinimidyl esters for coupling, biotinylation and conjugation with fluorophores. Further, it acts as a hydrophilic ligand and also used in covalently binding proteins to carboxyl containing beads, resins, other nanoparticles. It is involved in the conversion of carboxyl groups to primary amine-reactive sulfo-NHS esters.

Solubility
Soluble in methanol and water.

Notes
Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106627-54-7
Molecular Formula C4H4NNaO6S
Molecular Weight (g/mol) 217.127
MDL Number MFCD00043100
InChI Key RPENMORRBUTCPR-UHFFFAOYSA-M
Synonym n-hydroxysulfosuccinimide sodium salt, sulfo-nhs, sodium 1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate, n-hydroxysulfosuccinimidesodiumsalt, 3-pyrrolidinesulfonic acid, 1-hydroxy-2,5-dioxo-, monosodium salt, sodium 1-hydroxy-2,5-dioxo-pyrrolidine-3-sulfonate, n-hydroxusulfosuccinimide sodium, n-hydroxy-2-sodiosulfo succinimide, n-hydroxysulfosuccinimide,sodium salt, n-hydroxusulfosuccinimide sodium salt
PubChem CID 3520574
IUPAC Name sodium;1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate
SMILES C1C(C(=O)N(C1=O)O)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 250°C
Quantity 250 mg
Beilstein 6359129
Solubility Information Soluble in methanol and water.
Formula Weight 217.13
Percent Purity 97%
Chemical Name or Material N-Hydroxysulfosuccinimide sodium salt
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.