Learn More
N-Hydroxysulfosuccinimide sodium salt, 97%
Description
N-Hydroxysulfosuccinimide sodium salt is used as crosslinking agents for preparing hydrophilic active esters. It is also used to activate carboxylic acids to the more reactive succinimidyl esters for coupling, biotinylation and conjugation with fluorophores. Further, it acts as a hydrophilic ligand and also used in covalently binding proteins to carboxyl containing beads, resins, other nanoparticles. It is involved in the conversion of carboxyl groups to primary amine-reactive sulfo-NHS esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Hydroxysulfosuccinimide sodium salt is used as crosslinking agents for preparing hydrophilic active esters. It is also used to activate carboxylic acids to the more reactive succinimidyl esters for coupling, biotinylation and conjugation with fluorophores. Further, it acts as a hydrophilic ligand and also used in covalently binding proteins to carboxyl containing beads, resins, other nanoparticles. It is involved in the conversion of carboxyl groups to primary amine-reactive sulfo-NHS esters.
Solubility
Soluble in methanol and water.
Notes
Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 250°C |
| Quantity | 5 g |
| Beilstein | 6359129 |
| Solubility Information | Soluble in methanol and water. |
| Formula Weight | 217.13 |
| Percent Purity | 97% |
| Chemical Name or Material | N-Hydroxysulfosuccinimide sodium salt |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.