Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Iodosaccharin 98.0+%, TCI America™
SDP

Catalog No. I07845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07845G Supplier TCI America Supplier No. I07845G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86340-94-5
Molecular Formula C7H4INO3S
Molecular Weight (g/mol) 309.08
MDL Number MFCD09037939
InChI Key XQKQROJYWLWDMP-UHFFFAOYSA-N
Synonym n-iodosaccharin, unii-11wjr74obi, 11wjr74obi, 1,2-benzisothiazol-3 2h-one, 2-iodo-, 1,1-dioxide, 2-iodo-1,2-benzisothiazol-3 2h-one 1,1-dioxide, n-iodosaccharin mi, 2-iodo-1??,2-benzothiazole-1,1,3-trione, n-yodosaccharin, 2-iodobenzo d isothiazol-3 2h-one 1,1-dioxide
PubChem CID 8063362
IUPAC Name 2-iodo-2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
SMILES IN1C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

Quantity 5 g
Formula Weight 309.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Iodosaccharin
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.