N-Iodosuccinimide, 97%, Alfa Aesar™

Manufacturer: Alfa Aesar A1432006

Quantity 5g

 View more versions of this product

Catalog No. AAA1432006

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range 97%
    Molecular Formula C4H4INO2
    Formula Weight 224.99
    Chemical Name or Material N-Iodosuccinimide

    Specifications

    Formula Weight 224.99
    Quantity 5g
    Density 2.245
    Assay Percent Range 97%
    Molecular Formula C4H4INO2
    CAS 516-12-1
    Chemical Name or Material N-Iodosuccinimide

    Description

    Applications
    N-Iodosuccinimide is used in the preparation of vinyl sulfones from olefins and benzenesulfinic acid. It acts as a source for I+ and involved in Hunsdiecker reactions for the conversion of cinnamic acids, and propiolic acids to the corresponding alfa-halostyrenes and 1-halo-1-alkynes respectively. It is also used to hydrolyze thioglycosides to 1-hydroxyglycosides with triluoroacetic acid. It is involved in the preparation of iodobenzene from 1,6-diynes. Further, it acts as an iodinating agent in chemical synthesis.

    Solubility
    Soluble in dioxane, tetrahydrfuran and acetonitrile. Insoluble in ether and carbon tetrachloride.

    Notes
    Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit